missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Apolipoprotein A-IV Protein
A DNA sequence encoding the Mus musculus (Mouse) Apolipoprotein A-IV, was expressed in the hosts and tags indicated.
245.00 € - 2645.00 €
Specifications
Accession Number | NP_031494.2 |
---|---|
Gene ID (Entrez) | 11808 |
Name | Apolipoprotein A-IV Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986809
|
enQuireBio™
QP5662-YE-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926819
|
enQuireBio™
QP5662-YE-500UG |
500 μg |
2100.00 €
500µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996809
|
enQuireBio™
QP5662-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15916819
|
enQuireBio™
QP5662-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15936819
|
enQuireBio™
QP5662-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15906819
|
enQuireBio™
QP5662-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031494.2 | |
Apolipoprotein A-IV Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
11808 | |
Research Use Only | |
APOA4 | |
Recombinant Protein | |
Yeast | |
EVTSDQVANV VWDYFTQLSN NAKEAVEQFQ KTDVTQQLST LFQDKLGDAS TYADGVHNKL VPFVVQLSGH LAQETERVKE EIKKELEDLR DRMMPHANKV TQTFGENMQK LQEHLKPYAV DLQDQINTQT QEMKLQLTPY IQRMQTTIKE NVDNLHTSMM PLATNLKDKF NRNMEELKGH LTPRANELKA TIDQNLEDLR RSLAPLTVGV QEKLNHQMEG LAFQMKKNAE ELQTKVSAKI DQLQKNLAPL VEDVQSKVKG NTEGLQKSLE DLNRQLEQQV EEFRRTVEPM GEMFNKALVQ QLEQFRQQLG PNSGEVESHL SFLEKSLREK VNSFMSTLEK KGSPDQPQAL PLPEQAQEQA QEQAQEQVQP KPLES | |
Greater than 90% as determined by SDS-PAGE. |