missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ASGPR1 / ASGR1 Protein
A DNA sequence encoding the Mus musculus (Mouse) ASGPR1 / ASGR1, was expressed in the hosts and tags indicated.
245.00 € - 2645.00 €
Specifications
Accession Number | NP_001278060.1 |
---|---|
Gene ID (Entrez) | 11889 |
Name | ASGPR1 / ASGR1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986989
|
enQuireBio™
QP5684-YE-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926999
|
enQuireBio™
QP5684-YE-500UG |
500 μg |
2100.00 €
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996989
|
enQuireBio™
QP5684-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15916999
|
enQuireBio™
QP5684-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15936999
|
enQuireBio™
QP5684-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15906999
|
enQuireBio™
QP5684-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001278060.1 | |
ASGPR1 / ASGR1 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
11889 | |
Research Use Only | |
Asgr1 | |
Recombinant Protein | |
Yeast | |
QNSQLREDLL ALRQNFSNLT VSTEDQVKAL STQGSSVGRK MKLVESKLEK QQKDLTEDHS SLLLHVKQLV SDVRSLSCQM AAFRGNGSER TCCPINWVEY EGSCYWFSSS VRPWTEADKY CQLENAHLVV VTSRDEQNFL QRHMGPLNTW IGLTDQNGPW KWVDGTDYET GFQNWRPEQP DNWYGHGLGG GEDCAHFTTD GRWNDDVCRR PYRWVCETKL DKAN | |
Greater than 90% as determined by SDS-PAGE. |