missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ATP synthase subunit beta, mitochondrial Protein

A DNA sequence encoding the Mus musculus (Mouse) ATP synthase subunit beta, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5694-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927069

  • 2990.00 € / 1mg
Estimated Shipment: 20-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_058054.2
ATP synthase subunit beta, mitochondrial Protein
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
11947
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Atp5b
Recombinant Protein
Yeast
AAQASAAPKA GTATGRIVAV IGAVVDVQFD EGLPPILNAL EVQGRDSRLV LEVAQHLGES TVRTIAMDGT EGLVRGQKVL DSGAPIKIPV GPETLGRIMN VIGEPIDERG PIKTKQFAPI HAEAPEFIEM SVEQEILVTG IKVVDLLAPY AKGGKIGLFG GAGVGKTVLI MELINNVAKA HGGYSVFAGV GERTREGNDL YHEMIESGVI NLKDATSKVA LVYGQMNEPP GARARVALTG LTVAEYFRDQ EGQDVLLFID NIFRFTQAGS EVSALLGRIP SAVGYQPTLA TDMGTMQERI TTTKKGSITS VQAIYVPADD LTDPAPATTF AHLDATTVLS RAIAELGIYP AVDPLDSTSR IMDPNIVGNE HYDVARGVQK ILQDYKSLQD IIAILGMDEL SEEDKLTVSR ARKIQRFLSQ PFQVAEVFTG HMGKLVPLKE TIKGFQQILA GEYDHLPEQA FYMVGPIEEA VAKADKLAEE HGS
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers