missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse BID Protein

Product Code. 15966338
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15966338

Brand: enQuireBio™ QP111612ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the BID was constructed and used to recombinantly synthesize the protein.

Specifications

Name BID Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MDSEVSNGSG LGAEHITDLL VFGFLQSSGC TRQELEVLGR ELPVQAYWEA DLEDELQTDG SQASRSFNQG RIEPDSESQE EIIHNIARHL AQIGDEMDHN IQPTLVRQLA AQFMNGSLSE EDKRNCLAKA LDEVKTAFPR DMENDKAMLI MTMLLAKKVA SHAPSLLRDV FHTTVNFINQ NLFSYVRNLV RNEMD
Buffer The Mouse BID protein solution contains 10mM Tris-HCl pH 8.0, 1mM EDTA and 250mM NaCl.
Purity or Quality Grade Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.