missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Carboxylesterase 1C Protein
A DNA sequence encoding the Mus musculus (Mouse) Carboxylesterase 1C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7188-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031980.2 | |
Carboxylesterase 1C Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
13884 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Ces1c | |
Recombinant Protein | |
Yeast | |
HSLLPPVVDT TQGKVLGKYI SLEGFEQPVA VFLGVPFAKP PLGSLRFAPP QPAEPWSFVK NATSYPPMCS QDAGWAKILS DMFSTEKEIL PLKISEDCLY LNIYSPADLT KSSQLPVMVW IHGGGLVIGG ASPYNGLALS AHENVVVVTI QYRLGIWGLF STGDEHSPGN WAHLDQLAAL RWVQDNIANF GGNPDSVTIF GESSGGISVS VLVLSPLGKD LFHRAISESG VVINTNVGKK NIQAVNEIIA TLSQCNDTSS AAMVQCLRQK TESELLEISG KLVQYNISLS TMIDGVVLPK APEEILAEKS FNTVPYIVGF NKQEFGWIIP MMLQNLLPEG KMNEETASLL LRRFHSELNI SESMIPAVIE QYLRGVDDPA KKSELILDMF GDIFFGIPAV LLSRSLRDAG VSTYMYEFRY RPSFVSDKRP QTVEGDHGDE IFFVFGAPLL KEGASEEETN LSKMVMKFWA NFARNGNPNG EGLPHWPEYD EQEGYLQIGA TTQQAQRLKA EEVAFWTELL AKNPPETDPT EH |