missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL2 / MCP-1 / MCP1 Protein
Recombinant Protein
Brand: enQuireBio™ QP5449-1mg
Detalles adicionales : Weight : 0.01000kg
Especificaciones
NP_035463.1 | |
CCL2 / MCP-1 / MCP1 Protein | |
Research Use Only | |
CCL2 | |
Recombinant Protein | |
E. coli | |
QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN | |
Greater than 95% as determined by SDS-PAGE. |
20296 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
THP-1 chemotaxis significantly induced by less than 100 ng/ml of Mouse CCL2 / MCP-1 / MCP1. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |