missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CCL22 / MDC Protein

Product Code. 15953338
Click to view available options
Quantity:
100 μg
20 μg
250 μg
5 μg
Unit Size:
100µg
20µg
250µg
5µg
This item is not returnable. View return policy

Product Code. 15953338

Brand: enQuireBio™ QP10276ec5ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mouse CCL22 / MDC, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_033163.1
Gene ID (Entrez) 20299
Name CCL22 / MDC Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol CCL22
Protein Length Partial (see sequence information for more details).
Biological Activity The biological activity determined by a chemotaxis bioassay using human activated lymphocytes is in a concentration range of 10-100 ng/ml.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence GPYGANVEDS ICCQDYIRHP LPSRLVKEFF WTSKSCRKPG VVLITVKNRD ICADPRQVWV KKLLHKLS
Buffer 0.2 um filtered PBS, pH 7.4, lyophilized
Purity or Quality Grade Greater than 97% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.