missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD20 / MS4A1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD20 / MS4A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9289-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031667.1 | |
CD20 / MS4A1 Protein | |
Research Use Only | |
MS4A1 | |
Recombinant Protein | |
Yeast | |
VIMSSLSLFA AISGIILSIM DILNMTLSHF LKMRRLELIQ TSKPYVDIYD CEPSNSSEKN SPSTQYCNSI QSVFLGILSA MLISAFFQKL VTAGIVENEW KRMCTRSKSN VVLLSAGEKN EQTIKMKEEI IELSGVSSQP KNEEEIEIIP VQEEEEEEAE INFPAPPQEQ ESLPVENEIA P | |
Greater than 90% as determined by SDS-PAGE. |
12482 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |