missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD20 / MS4A1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD20 / MS4A1, was expressed in the hosts and tags indicated.
245.00 € - 2645.00 €
Specifications
Accession Number | NP_031667.1 |
---|---|
Gene ID (Entrez) | 12482 |
Name | CD20 / MS4A1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098011
|
enQuireBio™
QP9289-YE-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038021
|
enQuireBio™
QP9289-YE-500UG |
500 μg |
2100.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008021
|
enQuireBio™
QP9289-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028021
|
enQuireBio™
QP9289-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048021
|
enQuireBio™
QP9289-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018021
|
enQuireBio™
QP9289-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031667.1 | |
CD20 / MS4A1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
12482 | |
Research Use Only | |
MS4A1 | |
Recombinant Protein | |
Yeast | |
VIMSSLSLFA AISGIILSIM DILNMTLSHF LKMRRLELIQ TSKPYVDIYD CEPSNSSEKN SPSTQYCNSI QSVFLGILSA MLISAFFQKL VTAGIVENEW KRMCTRSKSN VVLLSAGEKN EQTIKMKEEI IELSGVSSQP KNEEEIEIIP VQEEEEEEAE INFPAPPQEQ ESLPVENEIA P | |
Greater than 90% as determined by SDS-PAGE. |