missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD20 / MS4A1 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD20 / MS4A1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9289-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048021

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_031667.1
CD20 / MS4A1 Protein
Research Use Only
MS4A1
Recombinant Protein
Yeast
VIMSSLSLFA AISGIILSIM DILNMTLSHF LKMRRLELIQ TSKPYVDIYD CEPSNSSEKN SPSTQYCNSI QSVFLGILSA MLISAFFQKL VTAGIVENEW KRMCTRSKSN VVLLSAGEKN EQTIKMKEEI IELSGVSSQP KNEEEIEIIP VQEEEEEEAE INFPAPPQEQ ESLPVENEIA P
Greater than 90% as determined by SDS-PAGE.
12482
50 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers