missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD27 Ligand/TNFSF7 Protein

Product Code. 18177590
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18177590

Brand: R&D Systems™ 783CL050/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to stimulate mouse T cell proliferation in the presence of anti-CD3. The ED50 for this effect is 1.25-5.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q05A52
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 970
Molecular Weight (g/mol) M.W. (Observed): 30 kDa, reducing conditions; M.W. (theoretical): 19.2 kDa
Name CD27 Ligand/TNFSF7
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse CD27 Ligand/TNFSF7 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse CD27 Ligand (Gln47 - Pro195) (N-terminus) Accession # Q05A52 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd70
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein CD27 Ligand/TNFSF7/CD70
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.