missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CD30 Ligand/TNFSF8 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
50µg
Description
The Recombinant Mouse CD30 Ligand/TNFSF8 Protein is derived from NS0. The Recombinant Mouse CD30 Ligand/TNFSF8 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 21949 |
| Molecular Weight (g/mol) | 22 kDa |
| Name | CD30 Ligand/TNFSF8 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse CD30 Ligand/TNFSF8 protein, (N-terminus) MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse CD30 Ligand (Gln68-Asp239) Accession &Num; P32972 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction