missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD40 Ligand/TNFSF5 (HA-tag) Protein

Product Code. 18706277
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18706277 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18706277 Supplier R&D Systems™ Supplier No. 8230CL050/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using mouse splenic B cells in the presence of IL-4. Banchereau, J. et al. (1991) Science 251:70. The ED50 for this effect is 0.07-0.35 ng/mL in the presence of a cross-linking antibody, Mouse Anti-Hemagglutinin/HA Peptide Monoclonal Antibody (Catalog # MAB060 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27548
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 959
Molecular Weight (g/mol) M.W. (Observed): 24-29 kDa, reducing conditions; M.W. (theoretical): 22 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse CD40 Ligand/TNFSF5 protein HA (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Mouse CD40 Ligand/TNFSF5 (Met112-Leu260) (N-terminus) Accession # P27548 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd40lg
Specific Reactivity CD40 Ligand/TNFSF5
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining
Protein CD40 Ligand/TNFSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.