missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CD90/Thy1 Fc Chimera Protein

Product Code. 18768547
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18768547 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18768547 Supplier R&D Systems™ Supplier No. 7335CD050
  • ONLINE EXCLUSIVE PRICE
    419.48 €
    373.80 € / 50µg
    Save 45.68 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse CD90/Thy1 Fc Chimera Protein is derived from NS0. The Recombinant Mouse CD90/Thy1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 21838
Molecular Weight (g/mol) 40 kDa (monomer)
Name CD90/Thy1 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse CD90/Thy1 protein, (N-terminus) Mouse CD90/Thy1 (Gln20-Lys130) Accession &Num; P01831 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.