missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CL-K1/COLEC11 Protein

Product Code. 18338747
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18338747

Brand: R&D Systems™ 4687CL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse CL-K1/COLEC11 Protein is derived from NS0. The Recombinant Mouse CL-K1/COLEC11 Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 71693
Molecular Weight (g/mol) 27 kDa
Name CL-K1/COLEC11
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse CL-K1/COLEC11 protein Gln27-Leu272,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.