missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CNOT7 Protein

A cDNA sequence encoding the CNOT7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11468-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927358

  • 3915.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CNOT7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPAATVD HSQRICEVWA CNLDEEMKKI RQVIRKYNYV AMDTEFPGVV ARPIGEFRSN ADYQYQLLRC NVDLLKIIQL GLTFMNEQGE YPPGTSTWQF NFKFNLTEDM YAQDSIELLT TSGIQFKKHE EEGIETQYFA ELLMTSGVVL CEGVKWLSFH SGYDFGYLIK ILTNSNLPEE ELDFFEILRL FFPVIYDVKY LMKSCKNLKM FFEDHIDDAK YCGHLYGLGS GSSYVQNGTG NAYEEEASKQ S
Greater than 80.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
CNOT7 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers