missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cyclophilin A Protein
A cDNA sequence encoding the Cyclophilin A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11577-5ug
Additional Details : Weight : 0.01000kg
Specifications
268373 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVNPTVF FDITADDEPL GRVSFELFAD KVPKTAENFR ALSTGEKGFG YKGSSFHRII PGFMCQGGDF TRHNGTGGRS IYGEKFEDEN FILKHTGPGI LSMANAGPNT NGSQFFICTA KTEWLDGKHV VFGKVKEGMN IVEAMERFGS RNGKTSKKIT ISDCGQL | |
Greater than 95% as determined by SDS-PAGE. |
Cyclophilin A Protein | |
Research Use Only | |
Ppia | |
Mouse | |
His | |
Cyclophilin A protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |