missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse DcTRAILR1/TNFRSF23 Protein
Description
The Recombinant Mouse DcTRAIL R1/TNFRSF23 Protein is derived from NS0. The Recombinant Mouse DcTRAIL R1/TNFRSF23 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q9ER63 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 8718 |
| Molecular Weight (g/mol) | Observed M.W.: 23-30kDa, reducing conditions, Theoretical M.W.: 15kDa |
| Name | DcTRAILR1/TNFRSF23 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse DcTRAIL R1/TNFRSF23 protein Met30-Ser156,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Common Name | DcTRAIL R1/TNFRSF23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?