missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse DPP4 / CD26 Protein

Product Code. p-7155875
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 16069121

Brand: enQuireBio™ QP9975iv10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) DPP4 / CD26, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001153015.1
Gene ID (Entrez) 13482
Name DPP4 / CD26 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Dpp4
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His-SUMO
Sequence SKDEAAADSR RTYSLADYLK STFRVKSYSL WWVSDFEYLY KQENNILLLN AEHGNSSIFL ENSTFESFGY HSVSPDRLFV LLEYNYVKQW RHSYTASYNI YDVNKRQLIT EEKIPNNTQW ITWSPEGHKL AYVWKNDIYV KVEPHLPSHR ITSTGEENVI YNGITDWVYE EEVFGAYSAL WWSPNNTFLA YAQFNDTGVP LIEYSFYSDE SLQYPKTVWI PYPKAGAVNP TVKFFIVNID SLSSSSSAAP IQIPAPASVA RGDHYLCDVV WATEERISLQ WLRRIQNYSV MAICDYDKIN LTWNCPSEQQ HVEMSTTGWV GRFRPAEPHF TSDGSSFYKI ISDKDGYKHI CHFPKDKKDC TFITKGAWEV ISIEALTSDY LYYISNQYKE MPGGRNLYKI QLTDHTNVKC LSCDLNPERC QYYAVSFSKE AKYYQLGCWG PGLPLYTLHR STDHKELRVL EDNSALDRML QDVQMPSKKL DFIVLNETRF WYQMILPPHF DKSKKYPLLL DVYAGPCSQK ADASFRLNWA TYLASTENII VASFDGRGSG YQGDKIMHAI NRRLGTLEVE DQIEAARQFV KMGFVDSKRV AIWGWSYGGY VTSMVLGSGS GVFKCGIAVA PVSRWEYYDS VYTERYMGLP IPEDNLDHYR NSTVMSRAEH FKQVEYLLIH GTADDNVHFQ QSAQISKALV DAGVDFQAMW YTDEDHGIAS STAHQHIYSH MSHFLQQCFS LH
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.