missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ENO1 / Enolase 1 / alpha-enolase Protein
A DNA sequence encoding the Mus musculus (Mouse) ENO1 / Enolase 1 / alpha-enolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5984-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001020559.1 | |
ENO1 / Enolase 1 / alpha-enolase Protein | |
Research Use Only | |
ENO1 | |
Recombinant Protein | |
E. coli | |
SILRIHAREI FDSRGNPTVE VDLYTAKGLF RAAVPSGAST GIYEALELRD NDKTRFMGKG VSQAVEHINK TIAPALVSKK VNVVEQEKID KLMIEMDGTE NKSKFGANAI LGVSLAVCKA GAVEKGVPLY RHIADLAGNP EVILPVPAFN VINGGSHAGN KLAMQEFMIL PVGASSFREA MRIGAEVYHN LKNVIKEKYG KDATNVGDEG GFAPNILENK EALELLKTAI AKAGYTDQVV IGMDVAASEF YRSGKYDLDF KSPDDPSRYI TPDQLADLYK SFVQNYPVVS IEDPFDQDDW GAWQKFTASA GIQVVGDDLT VTNPKRIAKA ASEKSCNCLL LKVNQIGSVT ESLQACKLAQ SNGWGVMVSH RSGETEDTFI ADLVVGLCTG QIKTGAPCRS ERLAKYNQIL RIEEELGSKA KFAGRSFRNP LA | |
Greater than 90% as determined by SDS-PAGE. |
13806 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |