missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Epididymal-specific lipocalin-5 Protein

A DNA sequence encoding the Mus musculus (Mouse) Epididymal-specific lipocalin-5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7423-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048790

  • 735.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001036095.1
Epididymal-specific lipocalin-5 Protein
Research Use Only
Lcn5
Recombinant Protein
E. coli
TEAAVVKDFD VNKFLGFWYE IALASKMGAY GLAHKEEKMG AMVVELKENL LALTTTYYNE GHCVLEKVAA TQVDGSAKYK VTRISGEKEV VVVATDYMTY TVIDITSLVA GAVHRAMKLY SRSLDNNGEA LNNFQKIALK HGFSETDIHI LKHDLTCVNA LQSGQI
Greater than 90% as determined by SDS-PAGE.
13863
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers