missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse FKBP1A Protein
A cDNA sequence encoding the FKBP1A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11885-1mg
Additional Details : Weight : 0.01000kg
Specifications
14225 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Fkbp1a | |
Mouse | |
His | |
The FKBP1A solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |
FKBP1A Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMGVQVE TISPGDGRTF PKRGQTCVVH YTGMLEDGKK FDSSRDRNKP FKFTLGKQEV IRGWEEGVAQ MSVGQRAKLI ISSDYAYGAT GHPGIIPPHA TLVFDVELLK LE |