missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Frizzled-5 Fc Chimera Protein

Product Code. 18757817
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18757817 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18757817 Supplier R&D Systems™ Supplier No. 9060FZ050
  • ONLINE EXCLUSIVE PRICE
    428.68 €
    382.20 € / 50µg
    Save 46.48 €
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Frizzled-5 Fc Chimera Protein is derived from NS0. The Recombinant Mouse Frizzled-5 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9EQD0
For Use With (Application) Bioactivity
Gene ID (Entrez) 7855
Molecular Weight (g/mol) Observed M.W.: 54-64kDa, reducing conditions, Theoretical M.W.: 43kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Frizzled-5 protein, (N-terminus) Mouse Frizzled-5 (Ala27-Pro167) Accession &Num; Q9EQD0 IEGRMD Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias C2orf31, chromosome 2 open reading frame 31, DKFZP434E2135, frizzled (Drosophila) homolog 5, frizzled homolog 5 (Drosophila), frizzled-5, Frizzled5, Frz-5, FZ-5, FZD5, FzE5, HFZ5, MGC129692, seven-transmembrane receptor frizzled-5, Wnt receptor
Common Name Frizzled-5
Gene Symbol Fzd5
Biological Activity Measured by its ability to inhibit Wnt-3a-induced alkaline phosphatase production by MC3T3-E1 mouse preosteoblast cells. The ED50 for this effect is typically 10-60ng/mL in the presence of 5ng/mL of Recombinant Mouse Wnt-3a.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse Frizzled-5 (Ala27-Pro167) Accession # Q9EQD0, N-terminus; IEGRMD; Mouse IgG2a (Glu98-Lys330), C-terminus
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.