missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse G-CSF / CSF3 Protein
Recombinant Protein
Brand: enQuireBio™ QP5415-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_034101.1 | |
G-CSF / CSF3 Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 50 pg/mL in the NFS-60 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^7 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium citrate, pH 3.0 |
12985 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
CSF3 | |
Recombinant Protein | |
E. coli | |
MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA |