missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Galectin-3 (Hek293-expressed) Protein
Description
Measured by the ability of the immobilized protein to support the adhesion of D10.G4.1 mouse helper T cells. The ED50 for this effect is 1-5 μg/mL.
Specifications
Specifications
| Accession Number | NP_034835 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl, TCEP, PEG and Trehalose. |
| Gene ID (Entrez) | 3958 |
| Molecular Weight (g/mol) | M.W. (Observed): 27-36 kDa, reducing conditions; M.W. (theoretical): 27 kDa |
| Name | AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived mouse Galectin-3 protein Met1-Ile264 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?