missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Glucagon receptor Protein

A DNA sequence encoding the Mus musculus (Mouse) Glucagon receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6073-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929789

  • 1925.00 € / 500µg
Estimated Shipment: 20-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032127.2
Glucagon receptor Protein
500 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
14527
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Gcgr
Recombinant Protein
Yeast
AQVMDFLFEK WKLYSDQCHH NLSLLPPPTE LVCNRTFDKY SCWPDTPPNT TANISCPWYL PWYHKVQHRL VFKRCGPDGQ WVRGPRGQPW RNASQCQLDD EEIEVQKGVA KMYSSQQ
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers