missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GM-CSF / CSF2 Protein
Recombinant Protein
2585.00 € - 4205.00 €
Specifications
Accession Number | NP_034099.2 |
---|---|
Gene ID (Entrez) | 12981 |
Name | GM-CSF / CSF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986089
|
enQuireBio™
QP5416-500UG |
500 μg |
2585.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976089
|
enQuireBio™
QP5416-1MG |
1 mg |
4205.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_034099.2 | |
GM-CSF / CSF2 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 50 pg/mL in the FDCP-1 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^7 units/mg. | |
Mouse | |
Untagged | |
10 mM acetic acid |
12981 | |
Research Use Only | |
CSF2 | |
Recombinant Protein | |
E. coli | |
MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK | |
Greater than 95% as determined by SDS-PAGE. |