missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GSTM1 Protein

A cDNA sequence encoding the GSTM1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10658-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924548

  • 131.00 € / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GSTM1 Protein
Research Use Only
Specific activity is 3 units/mg, and is defined as the amount of enzyme that conjugate 1.0 u mole of 1-chloro-2,4-dinitrobenzene (CDNB) with reduced glutathione per minute at pH 6.5 at 25C.
Mouse
Untagged
The GSTM1 solution contains PBS pH 7.4 and 5mM glutathione.
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MPMILGYWNV RGLTHPIRML LEYTDSSYDE KRYTMGDAPD FDRSQWLNEK FKLGLDFPNL PYLIDGSHKI TQSNAILRYL ARKHHLDGET EEERIRADIV ENQVMDTRMQ LIMLCYNPDF EKQKPEFLKT IPEKMKLYSE FLGKRPWFAG DKVTYVDFLA YDILDQYRMF EPKCLDAFPN LRDFLARFEG LKKISAYMKS SRYIATPIFS KMAHWSNK
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers