missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Guanylate cyclase activator 2B Protein
A DNA sequence encoding the Mus musculus (Mouse) Guanylate cyclase activator 2B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6134-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032217.2 | |
Guanylate cyclase activator 2B Protein | |
Research Use Only | |
Guca2b | |
Recombinant Protein | |
Yeast | |
VYIKYHGFQV QLESVKKLNE LEEKEMSNPQ PRRSGLLPAV CHNPALPLDL QPVCASQEAA STFKALRTIA TDECELCINV ACTGC | |
Greater than 90% as determined by SDS-PAGE. |
14916 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |