missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Guanylate cyclase activator 2B Protein

A DNA sequence encoding the Mus musculus (Mouse) Guanylate cyclase activator 2B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6134-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051050

  • 3260.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032217.2
Guanylate cyclase activator 2B Protein
Research Use Only
Guca2b
Recombinant Protein
E. coli
VYIKYHGFQV QLESVKKLNE LEEKEMSNPQ PRRSGLLPAV CHNPALPLDL QPVCASQEAA STFKALRTIA TDECELCINV ACTGC
Greater than 90% as determined by SDS-PAGE.
14916
1 mg
Not determined.
Full Length
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers