missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HAPLN1 Protein
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
Specifications
Specifications
| Accession Number | NP_038528.3 |
| Gene ID (Entrez) | 12950 |
| Name | HAPLN1 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not determined. |
| Gene Symbol | Hapln1 |
| Protein Length | Full Length |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction