missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IFN-alpha 2/IFNA2 Protein

Product Code. 18776896
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18776896 10 μg 10µg
18736276 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 18776896 Proveedor R&D Systems™ N.º de proveedor 10149IF010
  • ONLINE EXCLUSIVE PRICE
    338.64 €
    301.35 € / 10µg
    Save 37.29 €
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Interferon-alpha 2 (IFN alpha -2) is one of 14 subtypes within the IFN-alpha class of Type I Interferons

  • IFN-alpha class members signal through a common cell surface receptor complex composed of IFN-alpha R2 and IFN-alpha R1 subunits
  • The mature extracellular domain (ECD) of mouse IFN alpha -2 shares 60% and 83% aa sequence identity with human and rat, respectively
  • Murine IFN-alpha 2 can eliminate cardiac viral load and protect cardiomyocytes from injury in animals infected with coxsackievirus B3 (CVB3)
  • IFN alpha -2 derived mutants with reduced IFNR2 binding inhibited HIV replication and mutants with more IFNAR1 binding potentiated antiviral activity
TRUSTED_SUSTAINABILITY

Especificaciones

Accession Number P01573
For Use With (Application) Bioactivity
Gene ID (Entrez) 3440
Molecular Weight (g/mol) 19.3 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 2/IFNA2 protein Cys24-Glu190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after opening. 3 months, -20 to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Product Line R&D Systems
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.