missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IGFBP1 Protein

A DNA sequence encoding the Mus musculus (Mouse) IGFBP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6208-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051570

  • 735.00 € / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032367.3
IGFBP1 Protein
Research Use Only
Igfbp1
Recombinant Protein
E. coli
APQPWHCAPC TAERLGLCPP VPASCPEISR PAGCGCCPTC ALPMGAACGV ATARCAQGLS CRALPGEPRP LHALTRGQGA CVPEPAAPAT STLFSSQHEE AKAAVVSADE LSESPEMTEE QLLDSFHLMA PSREDQPILW NAISTYSSMR AREIADLKKW KEPCQRELYK VLERLAAAQQ KAGDEIYKFY LPNCNKNGFY HSKQCETSLD GEAGLCWCVY PWSGKKIPGS LETRGDPNCH QYFNVHN
Greater than 90% as determined by SDS-PAGE.
16006
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers