missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-12 Protein

Product Code. 18775977
Change view
Click to view available options
Quantity:
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation Unit Size
18775977 500 μg Lyophilized from a 0.2μm filtered solution in PBS. 500µg
18108150 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10µg
18375597 10 μg Lyophilized from a 0.2μm filtered solution in PBS. 10µg
18359677 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50µg
18108070 50 μg Lyophilized from a 0.2μm filtered solution in PBS. 50µg
18795879 500 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 18775977 Supplier R&D Systems™ Supplier No. 419ML500/CF
  • ONLINE EXCLUSIVE PRICE
    6936.67 €
    6231.75 € / 500µg
    Save 704.92 €
    10% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-12 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse IL-12 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 16159
Name IL-12
Quantity 500 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse IL-12 protein, (N-terminus) Mouse IL-12 p40 (Met23-Ser335) Accession &Num; P43432 Mouse IL-12 p35 (Arg23-Ala215) Accession &Num; NP_032377 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.