missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-17A (CHO-expressed) Protein

Product Code. 18716696
Change view
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18716696 10 μg 10µg
18746966 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18716696 Supplier R&D Systems™ Supplier No. 7956ML010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. ED50 for this effect is 0.25 to 1.25 ng/mL.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q62386
For Use With (Application) Bioactivity
Gene ID (Entrez) 3605
Molecular Weight (g/mol) 15 kDa (monomer)
Name CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse IL-17/IL-17A protein Ala26-Ala158
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol Il17a
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.