missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL 20 Protein

Product Code. 15934908
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15934908

Brand: enQuireBio™ QP107011mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IL 20 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 58181
Name IL 20 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol Il20
Biological Activity The biological activity was determined by measuring the dose dependent activation of STAT3 in human epidermal keratinocytes (HEK2a cells). Significant STAT3 activation is observed with >100ng/mL of Interleukin-20 recombinant mouse, corresponding to a specific activity of 10,000 units/mg of Interleukin-20 recombinant mouse.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MKGFGLAFGL FSAVGFLLWT PLTGLKTLHL GSCVITANLQ AIQKEFSEIR DSVQAEDTNI DIRILRTTES LKDIKSLDRC CFLRHLVRFY LDRVFKVYQT PDHHTLRKIS SLANSFLIIK KDLSVCHSHM ACHCGEEAME KYNQILSHFI ELELQAAVVK ALGELGILLR WMEEML
Buffer Lyophilized from a concentrated (1 mg/ml) solution containing no additives.
Purity or Quality Grade Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.