missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL 20 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Spezifikation
Spezifikation
| Gene ID (Entrez) | 58181 |
| Name | IL 20 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | Il20 |
| Biological Activity | The biological activity was determined by measuring the dose dependent activation of STAT3 in human epidermal keratinocytes (HEK2a cells). Significant STAT3 activation is observed with >100ng/mL of Interleukin-20 recombinant mouse, corresponding to a specific activity of 10,000 units/mg of Interleukin-20 recombinant mouse. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Species | E. coli |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur