missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-4 Protein
Description
The Recombinant Mouse IL-4 Protein is derived from E. coli. The Recombinant Mouse IL-4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P07750 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 3565 |
| Molecular Weight (g/mol) | 14 kDa |
| Quantity | 100 μg |
| Source | <EM>E. coli</EM>-derived mouse IL-4 protein His23-Ser140, with an N-terminal Met |
| Storage Requirements | Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?