missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL19 / Interleukin-19 Protein
Recombinant Protein
Brand: enQuireBio™ QP5483-1mg
Specifications
NP_001009940.1 | |
IL19 / Interleukin-19 Protein | |
Research Use Only | |
IL19 | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
329244 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MLRRCLISVD MRLIEKSFHE IKRAMQTKDT FKNVTILSLE NLRSIKPGDV CCMTNNLLTF YRDRVFQDHQ ERSLEVLRRI SSIANSFLCV QKSLERCQVH RQCNCSQEAT NATRIIHDNY NQLEVSSAAL KSLGELNILL AWIDRNHLET PAA | |
Greater than 95% as determined by SDS-PAGE. |