missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL7 / Interleukin 7 Protein
Recombinant Protein
Brand: enQuireBio™ QP5401-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032397.1 | |
IL7 / Interleukin 7 Protein | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MECHIKDKEG KAYESVLMIS IDELDKMTGT DSNCPNNEPN FFRKHVCDDT KEAAFLNRAA RKLKQFLKMN ISEEFNVHLL TVSQGTQTLV NCTSKEEKNV KEQKKNDACF LKRLLREIKT CWNKILKGSI |
16196 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
IL7 | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |