missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Leptin tA Protein

Product Code. p-7148573
Click to view available options
Quantity:
20 μg
5 μg
Unit Size:
20µg
5µg
This item is not returnable. View return policy

Product Code. 15945098

Brand: enQuireBio™ QP107645ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Leptin tA was constructed and used to recombinantly synthesize the protein.

Specifications

Name Leptin tA Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Leptin Antagonist Triple Mutant Mouse Recombinant half-life in circulation after SC injection was over 20 hours. Leptin Antagonist Triple Mutant Mouse Recombinant is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Leptin Antagonist Triple Mutant Mouse Recombinant in vitro activity is 5-6 fold lower than the non-pegylated antagonist, though in vivo it has profound weight gain effect (as compared to the non-pegylated antagonist), resulting mainly from increased food intake.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Buffer The Mouse Leptin triple antagonist was lyophilized from a concentrated (0.65 mg/ml) solution with 0.003mM NaHCO3.
Purity or Quality Grade Greater than 99.0% as determined by:(a) Gel filtration analysis. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.