missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LIF Protein
Recombinant Protein
Brand: enQuireBio™ QP4235-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001034626.1 | |
LIF Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the Inhibit M1 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
16878 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
LIF | |
Recombinant Protein | |
E. coli | |
MSPLPITPVN ATCAIRHPCH GNLMNQIKNQ LAQLNGSANA LFISYYTAQG EPFPNNVEKL CAPNMTDFPS FHGNGTEKTK LVELYRMVAY LSASLTNITR DQKVLNPTAV SLQVKLNATI DVMRGLLSNV LCRLCNKYRV GHVDVPPVPD HSDKEAFQRK KLGCQLLGTY KQVISVVVQA F |