missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MESDC2 Protein
A cDNA sequence encoding the MESDC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12681-10ug
Additional Details : Weight : 0.01000kg
Specifications
67943 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSADTPGEA TPPPRKKKDI RDYNDADMAR LLEQWEKDDD IEEGDLPEHK RPSAPIDFSK LDPGKPESIL KMTKKGKTLM MFVTVSGNPT EKETEEITSL WQGSLFNANY DVQRFIVGSD RAIFMLRDGS YAWEIKDFLV SQDRCAEVTL EGQMYPGKGG GSKEKNKTKP EKAKKKEGDP KPRASKEDNR AGSRREDL | |
Greater than 90.0% as determined by SDS-PAGE. |
MESDC2 Protein | |
Research Use Only | |
Mesd | |
Mouse | |
His | |
MESDC2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |