missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Methylmalonyl-CoA mutase, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Methylmalonyl-CoA mutase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6396-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032676.2 | |
Methylmalonyl-CoA mutase, mitochondrial Protein | |
Research Use Only | |
Mut | |
Recombinant Protein | |
E. coli | |
LHQQQPLHPE WAVLAKKQLK GKNPEDLIWH TPEGISIKPL YSRADTLDLP EELPGVKPFT RGPYPTMYTY RPWTIRQYAG FSTVEESNKF YKDNIKAGQQ GLSVAFDLAT HRGYDSDNPR VRGDVGMAGV AIDTVEDTKI LFDGIPLEKM SVSMTMNGAV IPVLATFIVT GEEQGVPKEK LTGTIQNDIL KEFMVRNTYI FPPEPSMKII ADIFQYTAQH MPKFNSISIS GYHMQEAGAD AILELAYTIA DGLEYCRTGL QAGLTIDEFA PRLSFFWGIG MNFYMEIAKM RAGRRLWAHL IEKMFQPKNS KSLLLRAHCQ TSGWSLTEQD PYNNIVRTAI EAMAAVFGGT QSLHTNSFDE ALGLPTVKSA RIARNTQIII QEESGIPKVA DPWGGSYMME SLTNDVYEAA LKLIYEVEEM GGMAKAVAEG IPKLRIEECA ARRQARIDSG SEVIVGVNKY QLEKEDSVEV LAIDNTSVRK KQIEKLKKIK SSRDQALAEQ CLSALTQCAA SGDGNILALA VDAARARCTV GEITDALKKV FGEHKANDRM VSGAYRQEFG ESKEITSAIK RVNKFMEREG RRPRLLVAKM GQDGHDRGAK VIATGFADLG FDVDIGPLFQ TPREVAQQAV DADVHAVGVS TLAAGHKTLV PELIKELTAL GRPDILVMCG GVIPPQDYEF LYEVGVSNVF GPGTRIPRAA VQVLDDIEKC LAEKQQSV | |
Greater than 90% as determined by SDS-PAGE. |
17850 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |