missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Methylmalonyl-CoA mutase, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Methylmalonyl-CoA mutase, mitochondrial, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_032676.2 |
---|---|
Gene ID (Entrez) | 17850 |
Name | Methylmalonyl-CoA mutase, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043020
|
enQuireBio™
QP6396-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063020
|
enQuireBio™
QP6396-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053020
|
enQuireBio™
QP6396-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073020
|
enQuireBio™
QP6396-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083020
|
enQuireBio™
QP6396-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093020
|
enQuireBio™
QP6396-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032676.2 | |
Methylmalonyl-CoA mutase, mitochondrial Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17850 | |
Research Use Only | |
Mut | |
Recombinant Protein | |
E. coli | |
LHQQQPLHPE WAVLAKKQLK GKNPEDLIWH TPEGISIKPL YSRADTLDLP EELPGVKPFT RGPYPTMYTY RPWTIRQYAG FSTVEESNKF YKDNIKAGQQ GLSVAFDLAT HRGYDSDNPR VRGDVGMAGV AIDTVEDTKI LFDGIPLEKM SVSMTMNGAV IPVLATFIVT GEEQGVPKEK LTGTIQNDIL KEFMVRNTYI FPPEPSMKII ADIFQYTAQH MPKFNSISIS GYHMQEAGAD AILELAYTIA DGLEYCRTGL QAGLTIDEFA PRLSFFWGIG MNFYMEIAKM RAGRRLWAHL IEKMFQPKNS KSLLLRAHCQ TSGWSLTEQD PYNNIVRTAI EAMAAVFGGT QSLHTNSFDE ALGLPTVKSA RIARNTQIII QEESGIPKVA DPWGGSYMME SLTNDVYEAA LKLIYEVEEM GGMAKAVAEG IPKLRIEECA ARRQARIDSG SEVIVGVNKY QLEKEDSVEV LAIDNTSVRK KQIEKLKKIK SSRDQALAEQ CLSALTQCAA SGDGNILALA VDAARARCTV GEITDALKKV FGEHKANDRM VSGAYRQEFG ESKEITSAIK RVNKFMEREG RRPRLLVAKM GQDGHDRGAK VIATGFADLG FDVDIGPLFQ TPREVAQQAV DADVHAVGVS TLAAGHKTLV PELIKELTAL GRPDILVMCG GVIPPQDYEF LYEVGVSNVF GPGTRIPRAA VQVLDDIEKC LAEKQQSV | |
Greater than 90% as determined by SDS-PAGE. |