missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MIP-3 beta / CCL19 Protein
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
Specifications
Specifications
| Name | MIP-3 beta / CCL19 Protein |
| Quantity | 500 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | Scya19 |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | GANDAEDCCL SVTQRPIPGN IVKAFRYLLN EDGCRVPAVV FTTLRGYQLC APPDQPWVDR IIRRLKKSSA KNKGNSTRRS PVS |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction