missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Muellerian-inhibiting factor Protein

A DNA sequence encoding the Mus musculus (Mouse) Muellerian-inhibiting factor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8974-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047351

  • 2100.00 € / 500µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Muellerian-inhibiting factor Protein
Research Use Only
AMH
Recombinant Protein
Yeast
DKGQDGPCAL RELSVDLRAE RSVLIPETYQ ANNCQGACRW PQSDRNPRYG NHVVLLLKMQ ARGAALGRLP CCVPTAYAGK LLISLSEERI SADHVPNMVA TEC
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions