missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nmes1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nmes1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7786-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040791

  • 1620.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001004174.1
Nmes1 Protein
Research Use Only
Nmes1
Recombinant Protein
Mammal
MGVFQILMKN KELIPLAFFI SVAATGATSF ALYALKKTDV VIDRKRNPEP WEMVDPTQPQ KLITINQQWK PVEELQKVRR ATR
Greater than 90% as determined by SDS-PAGE.
433470
100 μg
Not determined.
Full Length
Mouse
Flag
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers