missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NMNAT1 Protein
A cDNA sequence encoding the NMNAT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12858-50ug
Additional Details : Weight : 0.01000kg
Specifications
NMNAT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDSSKKT EVVLLACGSF NPITNMHLRL FELAKDYMHA TGKYSVIKGI ISPVGDAYKK KGLIPAHHRI IMAELATKNS HWVEVDTWES LQKEWVETVK VLRYHQEKLA TGSCSYPQSS PALEKPGRKR KWADQKQDSS PQKPQEPKPT GVPKVKLLCG ITNDISSTKI RRALRRGQSI RYLVPDLVQE YIEKHELYNT ESEGRNAGVT LAPLQRNAAE AKHNHSTL | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
NMNAT1 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM EDTA. |