missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse P4HB Protein
A cDNA sequence encoding the P4HB was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 18453 |
---|---|
Name | P4HB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | P4hb |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15902049
|
enQuireBio™
QP12948-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982039
|
enQuireBio™
QP12948-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15992039
|
enQuireBio™
QP12948-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
18453 | |
Research Use Only | |
P4hb | |
Mouse | |
His | |
P4HB protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
P4HB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DALEEEDNVL VLKKSNFEEA LAAHKYLLVE FYAPWCGHCK ALAPEYAKAA AKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFKN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLSDTAAAE SLVDSSEVTV IGFFKDVESD SAKQFLLAAE AIDDIPFGIT SNSGVFSKYQ LDKDGVVLFK KFDEGRNNFE GEITKEKLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLSSFKR AAEGFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESDELTAE KITEFCHRFL EGKIKPHLMS QEVPEDWDKQ PVKVLVGANF EEVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIIIAKMDS TANEVEAVKV HSFPTLKFFP ASADRTVIDY NGERTLDGFK KFLESGGQDG AGDDEDLDLE EALEPDMEED DDQKAVKDEL LEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |